missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hexafluorophosphate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Visit our Sensor Applications portal to learn more.

$189.76 - $642.58

Chemical Identifiers

MDL Number MFCD00011827
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014184
View Documents
Merck Emd Millipore
866185G
5 g
Each for $189.76
Only null left
 
111014183
View Documents
Merck Emd Millipore
8661825G
25 g
Each for $642.58
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00011827

Specifications

≥300°C
(C2H5)4 N(BF4)
5 g
217.06
For electrochemical analysis
429-06-1
MFCD00011827
Acetonitrile: 0.1 g/mL, clear, colorless
≥99.0% (T); ≥99.0%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'