missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hexafluorophosphate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014183
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014183 25 g
111014184 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014183 Supplier Merck Emd Millipore Supplier No. 8661825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00011827

Specifications

Melting Point ≥300°C
CAS 429-06-1
Linear Formula (C2H5)4 N(BF4)
Quantity 25 g
Solubility Information Acetonitrile: 0.1 g/mL, clear, colorless
Formula Weight 217.06
Percent Purity ≥99.0% (T); ≥99.0%
Grade For electrochemical analysis