missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium hexafluorophosphate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | ≥300°C |
| CAS | 429-06-1 |
| Linear Formula | (C2H5)4 N(BF4) |
| Quantity | 5 g |
| Solubility Information | Acetonitrile: 0.1 g/mL, clear, colorless |
| Molecular Weight (g/mol) | 217.06 |
| Formula Weight | 217.06 |
| Percent Purity | ≥99.0% (T); ≥99.0% |
| Grade | For electrochemical analysis |
| Physical Form | Crystals |