missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Chloride, Monohydrate, Crystal, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T10661KGBL

 View more versions of this product

Catalog No. 18614353


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

68696-18-4
Poly Bottle
0.98
1
1 kg
Reagent

Chemical Identifiers

68696-18-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.