missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium Chloride, Monohydrate, Crystal, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Bottle |
| Quantity | 25 g |
| Percent Purity | 0.98 |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.