CAS RN 68696-18-4
IUPAC Name:
tetraethylazanium hydrate chloride
Synonyms:
tetraethylammonium hydrate chloride
Molecular Weight (g/mol):
183.72
Molecular Formula:
C8H22ClNO
InChi Key:
CALLTGJPWMIDPC-UHFFFAOYSA-M
SMILES:
O.[Cl-].CC[N+](CC)(CC)CC
1
–
2
of
2
results
1
Promotions Available
Tetraethylammonium chloride hydrate, 98%
Tetraethylammonium chloride hydrate, 98%
CAS: 68696-18-4 Molecular Formula: C8H22ClNO Molecular Weight (g/mol): 183.72 MDL Number: MFCD00149992 InChI Key: CALLTGJPWMIDPC-UHFFFAOYSA-M Synonym: tetraethylammonium chloride hydrate,tetraethylammonium chloride monohydrate,tetraethylazanium chloride hydrate,tetraethylammonium hydrate chloride,c8h20n.cl.h2o,tetraethylammoniumchloride hydrate,tetraethyl ammonium chloride hydrate,tetraethylammonium chloride hydrate, cryst,n,n,n-triethylethanaminium chloride-water 1/1/1,tetraethylammonium chloride, monohydrate PubChem CID: 16211205 IUPAC Name: tetraethylazanium;chloride;hydrate SMILES: CC[N+](CC)(CC)CC.O.[Cl-]
2
Promotions Available
Tetraethylammonium Chloride, Monohydrate, Crystal, 98%, Spectrum™ Chemical
Tetraethylammonium Chloride, Monohydrate, Crystal, 98%, Spectrum™ Chemical
CAS: 68696-18-4