missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyl sodium sulfate, 95%, Thermo Scientific™

$282.84

Chemical Identifiers

CAS 1191-50-0
Molecular Formula C14H29NaO4S
Molecular Weight (g/mol) 316.43
MDL Number MFCD00007468
InChI Key UPUIQOIQVMNQAP-UHFFFAOYSA-M
Synonym sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
PubChem CID 23665770
IUPAC Name sodium;tetradecyl sulfate
SMILES [Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC420260050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $282.84
 
AC420260250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1191-50-0
316.43
UPUIQOIQVMNQAP-UHFFFAOYSA-M
23665770
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
C14H29NaO4S
MFCD00007468
sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
sodium;tetradecyl sulfate

Specifications

1191-50-0
Authentic
C14H29NaO4S
MFCD00007468
UPUIQOIQVMNQAP-UHFFFAOYSA-M
sodium;tetradecyl sulfate
23665770
95%
94% min.
94% min
CH3(CH2)13OSO3Na
sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
316.43
316.44
Tetradecyl sodium sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 214-737-2

SDS
Documents

Documents

RUO – Research Use Only