Learn More
Tetradecyl sodium sulfate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 420260250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C14H29NaO4S | |
| MFCD00007468 | |
| sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate | |
| sodium;tetradecyl sulfate |
Specifications
| Tetradecyl sodium sulfate | |
| 94% min. | |
| 94% min | |
| C14H29NaO4S | |
| MFCD00007468 | |
| sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate | |
| [Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O | |
| 316.43 | |
| 316.44 |
| 1191-50-0 | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)13OSO3Na | |
| 25g | |
| UPUIQOIQVMNQAP-UHFFFAOYSA-M | |
| sodium;tetradecyl sulfate | |
| 23665770 | |
| 95% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 214-737-2
Promotions