missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tetracosane | |
| 51°C | |
| 250°C | |
| MFCD00009352 | |
| n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b | |
| CCCCCCCCCCCCCCCCCCCCCCCC | |
| 338.664 | |
| CHEBI:32936 | |
| ≥99.0% (GC) |
| 646-31-1 | |
| White | |
| C24H50 | |
| 25 g | |
| POOSGDOYLQNASK-UHFFFAOYSA-N | |
| tetracosane | |
| 12592 | |
| 338.66 | |
| Crystalline Lumps |
Chemical Identifiers
| 646-31-1 | |
| 338.664 | |
| POOSGDOYLQNASK-UHFFFAOYSA-N | |
| 12592 | |
| tetracosane |
| C24H50 | |
| MFCD00009352 | |
| n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b | |
| CHEBI:32936 | |
| CCCCCCCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0010
TSCA : Yes