CAS RN 646-31-1
Tetracosane, 99%
CAS: 646-31-1 Molecular Formula: C24H50 Molecular Weight (g/mol): 338.65 MDL Number: MFCD00009352 InChI Key: POOSGDOYLQNASK-UHFFFAOYSA-N Synonym: n-tetracosane,lignocerane,unii-yq5h1m1d7i,tricosane, methyl,yq5h1m1d7i,tetracosane, analytical standard,tetracosane,acmc-1b0gq,n-tetracosane 5g,ksc357o9b PubChem CID: 12592 ChEBI: CHEBI:32936 IUPAC Name: tetracosane SMILES: CCCCCCCCCCCCCCCCCCCCCCCC
n-Tetracosane, 99%
CAS: 646-31-1 Molecular Formula: C24H50 Molecular Weight (g/mol): 338.664 MDL Number: MFCD00009352 InChI Key: POOSGDOYLQNASK-UHFFFAOYSA-N Synonym: n-tetracosane,lignocerane,unii-yq5h1m1d7i,tricosane, methyl,yq5h1m1d7i,tetracosane, analytical standard,tetracosane,acmc-1b0gq,n-tetracosane 5g,ksc357o9b PubChem CID: 12592 ChEBI: CHEBI:32936 IUPAC Name: tetracosane SMILES: CCCCCCCCCCCCCCCCCCCCCCCC
Tetracosane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Tetracosane belongs to the group of saturated hydrocarbons, which is found in lubricant oils.
Tetracosane, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™
Tetracosane belongs to the group of saturated hydrocarbons, which is found in lubricant oils.
Tetracosane 99.0+%, TCI America™
CAS: 646-31-1 Molecular Formula: C24H50 Molecular Weight (g/mol): 338.664 MDL Number: MFCD00009352 InChI Key: POOSGDOYLQNASK-UHFFFAOYSA-N Synonym: n-tetracosane,lignocerane,unii-yq5h1m1d7i,tricosane, methyl,yq5h1m1d7i,tetracosane, analytical standard,tetracosane,acmc-1b0gq,n-tetracosane 5g,ksc357o9b PubChem CID: 12592 ChEBI: CHEBI:32936 IUPAC Name: tetracosane SMILES: CCCCCCCCCCCCCCCCCCCCCCCC
Tetracosane 99.0+%, TCI America™
CAS: 646-31-1 Molecular Formula: C24H50 Molecular Weight (g/mol): 338.664 MDL Number: MFCD00009352 InChI Key: POOSGDOYLQNASK-UHFFFAOYSA-N Synonym: n-tetracosane,lignocerane,unii-yq5h1m1d7i,tricosane, methyl,yq5h1m1d7i,tetracosane, analytical standard,tetracosane,acmc-1b0gq,n-tetracosane 5g,ksc357o9b PubChem CID: 12592 ChEBI: CHEBI:32936 IUPAC Name: tetracosane SMILES: CCCCCCCCCCCCCCCCCCCCCCCC