missing translation for 'onlineSavingsMsg'
Learn More

Tetracosane 99.0+%, TCI America™

Catalog No. p-7136065
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
T007525G 25 g
T0075100G 100 g
T0075500G 500 g
3 options

Catalog No. T007525G

Supplier: TCI America T007525G

Only null left

Chemical Identifiers

CAS 646-31-1
Molecular Formula C24H50
Molecular Weight (g/mol) 338.664
MDL Number MFCD00009352
InChI Key POOSGDOYLQNASK-UHFFFAOYSA-N
Synonym n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b
PubChem CID 12592
ChEBI CHEBI:32936
IUPAC Name tetracosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 51°C
Color White
Boiling Point 250°C
Quantity 25 g
Formula Weight 338.66
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tetracosane
EINECSNumber (2)-0010
TSCA Yes