missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorophthalic Acid Hemihydrate 98.0+%, TCI America™

Catalog No. T007025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T007025G 25 g
T0070500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T007025G Supplier TCI America Supplier No. T007025G

Chemical Identifiers

CAS 632-58-6
Molecular Formula C8H2Cl4O4
Molecular Weight (g/mol) 303.90
MDL Number MFCD00053309
InChI Key WZHHYIOUKQNLQM-UHFFFAOYSA-N
PubChem CID 12442
IUPAC Name tetrachlorobenzene-1,2-dicarboxylic acid
SMILES OC(=O)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 303.90
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrachlorophthalic Acid Hemihydrate
EINECSNumber (3)-0062
RTECSNumber TI2795000
TSCA Yes