accessibility menu, dialog, popup

UserName

CAS RN 632-58-6

HOOClClClClOHO

CAS RN 632-58-6

IUPAC Name: tetrachlorobenzene-1,2-dicarboxylic acid
Synonyms: tetrachlorophthalic acid
Molecular Weight (g/mol): 303.9
Molecular Formula: C8H2Cl4O4
InChi Key: WZHHYIOUKQNLQM-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results