missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachlorohydroquinone 98.0+%, TCI America™
Supplier: TCI America T006725G
Specifications
| Tetrachlorohydroquinone | |
| 242°C | |
| MFCD00041748 | |
| tetrachlorohydroquinone, 2,3,5,6-tetrachlorohydroquinone, dihydro-p-chloranil, tetrachloro-p-benzohydroquinone, tetrachlorobenzoquinol, nordrosophilin a, 2,3,5,6-tetrachloro-1,4-benzenediol, perchlorohydroquinone, hydroquinone, tetrachloro, 1,4-benzenediol, 2,3,5,6-tetrachloro | |
| C1(=C(C(=C(C(=C1Cl)Cl)O)Cl)Cl)O | |
| 247.88 | |
| CHEBI:17042 | |
| ≥98.0% (GC) |
| 87-87-6 | |
| C6H2Cl4O2 | |
| 25 g | |
| STOSPPMGXZPHKP-UHFFFAOYSA-N | |
| 2,3,5,6-tetrachlorobenzene-1,4-diol | |
| 66603 | |
| 247.88 | |
| Crystalline Powder |
Chemical Identifiers
| 87-87-6 | |
| 247.88 | |
| STOSPPMGXZPHKP-UHFFFAOYSA-N | |
| 66603 | |
| 2,3,5,6-tetrachlorobenzene-1,4-diol |
| C6H2Cl4O2 | |
| MFCD00041748 | |
| tetrachlorohydroquinone, 2,3,5,6-tetrachlorohydroquinone, dihydro-p-chloranil, tetrachloro-p-benzohydroquinone, tetrachlorobenzoquinol, nordrosophilin a, 2,3,5,6-tetrachloro-1,4-benzenediol, perchlorohydroquinone, hydroquinone, tetrachloro, 1,4-benzenediol, 2,3,5,6-tetrachloro | |
| CHEBI:17042 | |
| C1(=C(C(=C(C(=C1Cl)Cl)O)Cl)Cl)O |
Safety and Handling
RTECSNumber : MX7700000
TSCA : Yes