missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracaine 98.0+%, TCI America™
$83.89 - $244.19
Chemical Identifiers
| CAS | 94-24-6 |
|---|---|
| Molecular Formula | C15H24N2O2 |
| Molecular Weight (g/mol) | 264.369 |
| MDL Number | MFCD00053787 |
| InChI Key | GKCBAIGFKIBETG-UHFFFAOYSA-N |
| Synonym | 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester |
| PubChem CID | 5411 |
| ChEBI | CHEBI:9468 |
| IUPAC Name | 2-(dimethylamino)ethyl 4-(butylamino)benzoate |
| SMILES | CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C |
Chemical Identifiers
| 94-24-6 | |
| 264.369 | |
| GKCBAIGFKIBETG-UHFFFAOYSA-N | |
| 5411 | |
| 2-(dimethylamino)ethyl 4-(butylamino)benzoate |
| C15H24N2O2 | |
| MFCD00053787 | |
| 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester | |
| CHEBI:9468 | |
| CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C |
Specifications
| 94-24-6 | |
| White-Yellow | |
| MFCD00053787 | |
| 2811 | |
| GKCBAIGFKIBETG-UHFFFAOYSA-N | |
| 2-(dimethylamino)ethyl 4-(butylamino)benzoate | |
| 5411 | |
| 264.37 | |
| Crystalline Powder |
| 43°C | |
| C15H24N2O2 | |
| 5 g | |
| 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester | |
| CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C | |
| 264.369 | |
| CHEBI:9468 | |
| ≥98.0% (GC,T) | |
| Tetracaine |
Safety and Handling
RTECSNumber : DG4725000
TSCA : No
Recommended Storage : Refrigerator