missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine 98.0+%, TCI America™

Catalog No. t27895g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27895G 5 g
T278925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27895G Supplier TCI America Supplier No. T27895G
Only null left

Chemical Identifiers

CAS 94-24-6
Molecular Formula C15H24N2O2
Molecular Weight (g/mol) 264.369
MDL Number MFCD00053787
InChI Key GKCBAIGFKIBETG-UHFFFAOYSA-N
Synonym 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester
PubChem CID 5411
ChEBI CHEBI:9468
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate
SMILES CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 264.37
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetracaine
RTECSNumber DG4725000
TSCA No
Recommended Storage Refrigerator