missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine 98.0+%, TCI America™

Supplier:  TCI America T278925G

 View more versions of this product

Catalog No. T278925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetracaine
43°C
C15H24N2O2
25 g
2-(Dimethylamino)ethyl 4-(Butylamino)benzoate, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester
CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
264.369
CHEBI:9468
≥98.0% (GC,T)
94-24-6
White-Yellow
MFCD00053787
2811
GKCBAIGFKIBETG-UHFFFAOYSA-N
2-(dimethylamino)ethyl 4-(butylamino)benzoate
5411
264.37
Crystalline Powder

Chemical Identifiers

94-24-6
264.369
GKCBAIGFKIBETG-UHFFFAOYSA-N
5411
2-(dimethylamino)ethyl 4-(butylamino)benzoate
C15H24N2O2
MFCD00053787
2-(Dimethylamino)ethyl 4-(Butylamino)benzoate, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester
CHEBI:9468
CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DG4725000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions