missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracaine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 94-24-6 |
|---|---|
| Molecular Formula | C15H24N2O2 |
| Molecular Weight (g/mol) | 264.36 |
| InChI Key | GKCBAIGFKIBETG-UHFFFAOYSA-N |
| PubChem CID | 5411 |
| ChEBI | CHEBI:9468 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC460820050
|
Thermo Scientific Chemicals
460820050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC460820250
|
Thermo Scientific Chemicals
460820250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC460821000
|
Thermo Scientific Chemicals
460821000 |
100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Danger
EINECSNumber : 202-316-6
RUO â Research Use Only