missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydroxide Titrant, 0.4 M Solution in Water, Spectrum™ Chemical
$668.19 - $2,022.97
Chemical Identifiers
| CAS | 2052-49-5 |
|---|---|
| Molecular Formula | C16H37NO |
| Molecular Weight (g/mol) | 259.48 |
| MDL Number | MFCD00009425 |
| InChI Key | VDZOOKBUILJEDG-UHFFFAOYSA-M |
| IUPAC Name | tetrabutylazanium hydroxide |
| SMILES | [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18614345
|
Spectrum Chemical Mfg Cor
T2431100ML |
100 mL | Amber Glass Bottle |
N/A
|
|
||||
|
18614346
|
Spectrum Chemical Mfg Cor
T2431500ML |
500 mL | Amber Glass Bottle |
N/A
|
|
||||
|
18614347
|
Spectrum Chemical Mfg Cor
T24314LT |
4 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 2052-49-5 | |
| 259.48 | |
| VDZOOKBUILJEDG-UHFFFAOYSA-M | |
| [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
| C16H37NO | |
| MFCD00009425 | |
| tetrabutylazanium hydroxide |
Specifications
| 2052-49-5,7732-18-5 | |
| 1 | |
| C16H37NO | |
| 100 mL | |
| [OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
| 259.48 | |
| 0.37- 0.43 M |
| 0.15 | |
| Amber Glass Bottle | |
| MFCD00009425 | |
| VDZOOKBUILJEDG-UHFFFAOYSA-M | |
| tetrabutylazanium hydroxide | |
| Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.