missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydroxide Titrant, 0.4 M Solution in Water, Spectrum™ Chemical
Specifications
Specifications
| Absorbance | 0.15 |
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 4 L |
| Grade | Reagent |
| Concentration or Composition (by Analyte or Components) | 0.37- 0.43 M |
| Physical Form | Liquid |
| Chemical Name or Material | Tetrabutylammonium Hydroxide Titrant |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.