accessibility menu, dialog, popup

UserName

NH3CCH3H3CCH3OH

CAS RN 2052-49-5

IUPAC Name: tetrabutylazanium hydroxide
Synonyms: tetrabutylammonium hydroxide
Molecular Weight (g/mol): 259.48
Molecular Formula: C16H37NO
InChi Key: VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES: [OH-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (52)
Quantity 
  • (2)
  • (1)
  • (11)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (11)
  • (1)
Physical Form 
  • (4)
  • (28)
  • (7)
Grade 
  • (1)
  • (6)
  • (2)
  • (6)
  • (1)
Boiling Point 
  • (4)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (19)
  • (9)
  • (1)
  • (3)
  • (2)
  • (6)
  • (9)
  • (34)
  • (3)
  • (52)
  • (2)
  • (1)
  • (11)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (11)
  • (1)
  • (4)
  • (28)
  • (7)
  • (1)
  • (6)
  • (2)
  • (6)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)