missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide Solution, ∼40 in Water, MilliporeSigma™ Supelco™

Tetrabutylammonium hydroxide (TBAOH) is a quaternary ammonium salt,mainly used as a strong base and phase-transfer catalyst.

$274.63 - $3906.84

Chemical Identifiers

MDL Number MFCD00009425
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111018960
View Documents
Merck Emd Millipore
86854100ML
100 mL
Each for $274.63
Only null left
 
111017937
View Documents
Merck Emd Millipore
8685425L
2.5 L
Each for $3,906.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00009425

Specifications

2052-49-5
(CH3 CH2 CH2 CH2)4 N(OH)
259.47
40% aq. soln.
100 mL
MFCD00009425
Ion Chromatography
0.995 g/cm3
Videos
SDS
Documents

Documents