missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide Solution, ∼40 in Water, MilliporeSigma™ Supelco™

Catalog No. 111018960
Change view
Click to view available options
Quantity:
100 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018960 100 mL
111017937 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018960 Supplier Merck Emd Millipore Supplier No. 86854100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetrabutylammonium hydroxide (TBAOH) is a quaternary ammonium salt,mainly used as a strong base and phase-transfer catalyst.

Chemical Identifiers

MDL Number MFCD00009425

Specifications

CAS 2052-49-5
Physical Form Liquid
Quantity 100 mL
Linear Formula (CH3 CH2 CH2 CH2)4 N(OH)
Molecular Weight (g/mol) 259.47
Formula Weight 259.47
Grade Ion Chromatography
Concentration or Composition (by Analyte or Components) 40% aq. soln.
Packaging Poly Bottle
Density 0.995 g/cm3
Chemical Name or Material Tetrabutylammonium Hydroxide
Show More Show Less