missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydroxide, 1.0 M Solution in Methanol, Spectrum™ Chemical

Catalog No. 18614343
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614343 100 mL Amber Glass Bottle
18614342 25 mL Amber Glass Bottle
18614344 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18614343 Supplier Spectrum Chemical Mfg Cor Supplier No. T1058100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 2052-49-5 , 7732-18-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
IUPAC Name tetrabutylazanium hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

CAS Max % 1
Boiling Point 64°C
Packaging Amber Glass Bottle
Quantity 100 mL
Grade Reagent
Concentration or Composition (by Analyte or Components) 0.9-1.1 M
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Hydroxide in Methanol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.