missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydroxide, 1.0 M Solution in Methanol, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 64°C |
| Packaging | Amber Glass Bottle |
| Quantity | 25 mL |
| Grade | Reagent |
| Concentration or Composition (by Analyte or Components) | 0.9-1.1 M |
| Physical Form | Liquid |
| Chemical Name or Material | Tetrabutylammonium Hydroxide in Methanol |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.