missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydroxide, 0.10 N in Isopropanol/Methanol, Reagents

Catalog No. 01333466
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333466 1 L
01333465 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333466 Supplier Labchem Inc Supplier No. CT0060001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetrabutylammonium Hydroxide, 0.10 N in Isopropanol/Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 67-56-1 , 2052-49-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.01
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Hydroxide, 0.10 N in Isopropanol/Methanol