missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Fluoride (ca. 1mol/L in Tetrahydrofuran), TCI America™

Supplier:  TCI America T1338500ML

 View more versions of this product

Catalog No. T1338500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetrabutylammonium Fluoride (ca. 1mol/L in Tetrahydrofuran)
429-41-4
MFCD00011747
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
[F-].CCCC[N+](CCCC)(CCCC)CCCC
261.47
CHEBI:51990
500 mL
Undesignated
C16H36FN
2924
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylazanium fluoride
2724141
261.47
Clear Liquid at 20°C

Chemical Identifiers

429-41-4
261.47
FPGGTKZVZWFYPV-UHFFFAOYSA-M
2724141
tetrabutylazanium fluoride
C16H36FN
MFCD00011747
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
CHEBI:51990
[F-].CCCC[N+](CCCC)(CCCC)CCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions