missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Fluoride (ca. 1mol/L in Tetrahydrofuran), TCI America™
$53.16 - $349.34
Chemical Identifiers
| CAS | 429-41-4 |
|---|---|
| Molecular Formula | C16H36FN |
| Molecular Weight (g/mol) | 261.47 |
| MDL Number | MFCD00011747 |
| InChI Key | FPGGTKZVZWFYPV-UHFFFAOYSA-M |
| Synonym | tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 |
| PubChem CID | 2724141 |
| ChEBI | CHEBI:51990 |
| IUPAC Name | tetrabutylazanium fluoride |
| SMILES | [F-].CCCC[N+](CCCC)(CCCC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T133825ML
|
TCI America
T133825ML |
25 mL |
Each for $53.16
|
|
|||||
|
T1338100ML
|
TCI America
T1338100ML |
100 mL |
Each for $125.31
|
|
|||||
|
T1338500ML
|
TCI America
T1338500ML |
500 mL |
Each for $349.34
|
|
|||||
Chemical Identifiers
| 429-41-4 | |
| 261.47 | |
| FPGGTKZVZWFYPV-UHFFFAOYSA-M | |
| 2724141 | |
| tetrabutylazanium fluoride |
| C16H36FN | |
| MFCD00011747 | |
| tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 | |
| CHEBI:51990 | |
| [F-].CCCC[N+](CCCC)(CCCC)CCCC |
Specifications
| Undesignated | |
| C16H36FN | |
| 2924 | |
| FPGGTKZVZWFYPV-UHFFFAOYSA-M | |
| tetrabutylazanium fluoride | |
| 2724141 | |
| 261.47 | |
| Clear Liquid at 20°C |
| 429-41-4 | |
| MFCD00011747 | |
| tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 | |
| [F-].CCCC[N+](CCCC)(CCCC)CCCC | |
| 261.47 | |
| CHEBI:51990 | |
| 25 mL | |
| Tetrabutylammonium Fluoride (ca. 1mol/L in Tetrahydrofuran) |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes
Recommended Storage : Refrigerator