missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickelate 98.0+%, TCI America™
Supplier: TCI America T3220200MG
Specifications
| Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickelate | |
| 148°C | |
| C30H48NNiS4 | |
| 200 mg | |
| OGDOOYYRRAJVOI-UHFFFAOYSA-J | |
| λ³-nickel(3+) bis((5-methyl-2-sulfanidylphenyl)sulfanide) tetrabutylazanium | |
| 86280311 | |
| ≥98.0% (HPLC,T) |
| 15492-42-9 | |
| Black-Yellow | |
| MFCD00216666,MFCD12022530 | |
| Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickel(III) Complex | |
| [Ni+3].CC1=CC([S-])=C([S-])C=C1.CC1=CC([S-])=C([S-])C=C1.CCCC[N+](CCCC)(CCCC)CCCC | |
| 609.65 | |
| 609.65 | |
| Crystalline Powder |
Chemical Identifiers
| 15492-42-9 | |
| 609.65 | |
| OGDOOYYRRAJVOI-UHFFFAOYSA-J | |
| 86280311 | |
| [Ni+3].CC1=CC([S-])=C([S-])C=C1.CC1=CC([S-])=C([S-])C=C1.CCCC[N+](CCCC)(CCCC)CCCC |
| C30H48NNiS4 | |
| MFCD00216666,MFCD12022530 | |
| Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickel(III) Complex | |
| λ³-nickel(3+) bis((5-methyl-2-sulfanidylphenyl)sulfanide) tetrabutylazanium |
Safety and Handling
TSCA : No