missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickelate 98.0+%, TCI America™
$287.06 - $860.63
Chemical Identifiers
| CAS | 15492-42-9 |
|---|---|
| Molecular Formula | C30H48NNiS4 |
| Molecular Weight (g/mol) | 609.65 |
| MDL Number | MFCD00216666,MFCD12022530 |
| InChI Key | OGDOOYYRRAJVOI-UHFFFAOYSA-J |
| Synonym | Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickel(III) Complex |
| PubChem CID | 86280311 |
| IUPAC Name | λ³-nickel(3+) bis((5-methyl-2-sulfanidylphenyl)sulfanide) tetrabutylazanium |
| SMILES | [Ni+3].CC1=CC([S-])=C([S-])C=C1.CC1=CC([S-])=C([S-])C=C1.CCCC[N+](CCCC)(CCCC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3220200MG
|
TCI America
T3220200MG |
200 mg |
Each for $287.06
|
|
|||||
|
T32201G
|
TCI America
T32201G |
1 g |
Each for $860.63
|
|
|||||
Chemical Identifiers
| 15492-42-9 | |
| 609.65 | |
| OGDOOYYRRAJVOI-UHFFFAOYSA-J | |
| 86280311 | |
| [Ni+3].CC1=CC([S-])=C([S-])C=C1.CC1=CC([S-])=C([S-])C=C1.CCCC[N+](CCCC)(CCCC)CCCC |
| C30H48NNiS4 | |
| MFCD00216666,MFCD12022530 | |
| Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickel(III) Complex | |
| λ³-nickel(3+) bis((5-methyl-2-sulfanidylphenyl)sulfanide) tetrabutylazanium |
Specifications
| 15492-42-9 | |
| Black-Yellow | |
| MFCD00216666,MFCD12022530 | |
| Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickel(III) Complex | |
| [Ni+3].CC1=CC([S-])=C([S-])C=C1.CC1=CC([S-])=C([S-])C=C1.CCCC[N+](CCCC)(CCCC)CCCC | |
| 609.65 | |
| 609.65 | |
| Crystalline Powder |
| 148°C | |
| C30H48NNiS4 | |
| 200 mg | |
| OGDOOYYRRAJVOI-UHFFFAOYSA-J | |
| λ³-nickel(3+) bis((5-methyl-2-sulfanidylphenyl)sulfanide) tetrabutylazanium | |
| 86280311 | |
| ≥98.0% (HPLC,T) | |
| Tetrabutylammonium Bis(4-methyl-1,2-benzenedithiolato)nickelate |
Safety and Handling
TSCA : No