accessibility menu, dialog, popup

UserName

H3CNCH3H3CH3CCH3SSCH3SSNi

CAS RN 15492-42-9

IUPAC Name: λ³-nickel(3+) bis((5-methyl-2-sulfanidylphenyl)sulfanide) tetrabutylazanium
Synonyms: λ³-nickel(3+) bis((5-methyl-2-sulfanidylphenyl)sulfanide) tetrabutylammonium ion
Molecular Weight (g/mol): 609.65
Molecular Formula: C30H48NNiS4
InChi Key: OGDOOYYRRAJVOI-UHFFFAOYSA-J
SMILES: [Ni+3].CC1=CC([S-])=C([S-])C=C1.CC1=CC([S-])=C([S-])C=C1.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results