missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromobisphenol A Bis(2-hydroxyethyl) Ether 93.0+%, TCI America™

Catalog No. T0851500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0851500G 500 g
T085125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0851500G Supplier TCI America Supplier No. T0851500G
Only null left

Chemical Identifiers

CAS 4162-45-2
Molecular Formula C19H20Br4O4
Molecular Weight (g/mol) 631.981
MDL Number MFCD00059603
InChI Key RVHUMFJSCJBNGS-UHFFFAOYSA-N
Synonym 2,2-Bis[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]propane, O,O-Bis(2-hydroxyethyl)tetrabromobisphenol A
PubChem CID 20113
IUPAC Name 2-[2,6-dibromo-4-[2-[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]propan-2-yl]phenoxy]ethanol
SMILES CC(C)(C1=CC(=C(C(=C1)Br)OCCO)Br)C2=CC(=C(C(=C2)Br)OCCO)Br

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 500 g
Formula Weight 631.98
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromobisphenol A Bis(2-hydroxyethyl) Ether
EINECSNumber (4)-0218
TSCA Yes