missing translation for 'onlineSavingsMsg'
Learn More

TES sodium salt, Thermo Scientific Chemicals

A zwitterionic Good's Buffer

$102.92 - $102.92

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.23
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
Synonym 2-([Tris(hydroxymethyl)methyl]amino)ethane-1-sulfonic acid sodium salt
PubChem CID 23702152
IUPAC Name sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
SMILES [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6270614
View Documents
Thermo Scientific Chemicals
J6270614
25 g
Each for $102.92
Only null left
 
Description

Description

TES has potential for a variety of biological applications. In many culture media that require metal cations, many buffers cannot be used due to chelation or precipitation (citrate or phosphate, for example). TES was found to be beneficial in a study of succinate oxidation. TES is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9 where additional buffering capacity is required. It is also useful in cell culture media formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TES has potential for a variety of biological applications. In many culture media that require metal cations, many buffers cannot be used due to chelation or precipitation (citrate or phosphate, for example). TES was found to be beneficial in a study of succinate oxidation. TES is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9 where additional buffering capacity is required. It is also useful in cell culture media formulations.

Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Chemically stable, and stable to temperature and concentration changes.
Specifications

Chemical Identifiers

70331-82-7
251.23
GUXMFAHOYNRHBI-UHFFFAOYSA-M
23702152
[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
C6H14NNaO6S
MFCD00065482
2-([Tris(hydroxymethyl)methyl]amino)ethane-1-sulfonic acid sodium salt
sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate

Specifications

70331-82-7
Powder
C6H14NNaO6S
1957061
2-([Tris(hydroxymethyl)methyl]amino)ethane-1-sulfonic acid sodium salt
GUXMFAHOYNRHBI-UHFFFAOYSA-M
sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
23702152
TES sodium salt
White
25 g
MFCD00065482
14,9176
Soluble in water (350g/L) at 20°C,and methanol(very faint turbidity).
[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
251.23
251.23
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only