missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TES, 99%, for biochemistry

Catalog No. AC207510250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207510250 25 g Glass bottle
AC207511000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207510250 Supplier Thermo Scientific Chemicals Supplier No. 207510250

CAS: 7365-44-8 | C6H15NO6S | 229.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 7365-44-8
Molecular Formula C6H15NO6S
Molecular Weight (g/mol) 229.247
MDL Number MFCD00007532
InChI Key JOCBASBOOFNAJA-UHFFFAOYSA-N
Synonym 2-[Tris(hydroxymethyl)methylamino]-1-ethanesulfonic acid
PubChem CID 81831
ChEBI CHEBI:44356
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NC(CO)(CO)CO

Specifications

Absorbance (0.1M aq. soln.), (0.1M aq. soln.) at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 223.0°C to 225.0°C
Color White
pH 4 to 8 (0.1M soln. at 20°C)
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 1% max.
Linear Formula (HOCH2)3CNHCH2CH2SO3H
Packaging Glass bottle
Merck Index 15, 9319
Solubility Information Solubility in water: soluble.
Formula Weight 229.25
Percent Purity 99%
Grade Biochemical
Chemical Name or Material TES, For biochemistry
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only