missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TES, 0.2M buffer soln., pH 7.5

Catalog No. AAJ61350AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61350AK 250 mL
AAJ61350AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61350AK Supplier Thermo Scientific Chemicals Supplier No. J61350AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70331-82-7 | C6H14NNaO6S | 251.23 g/mol

TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.23
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
PubChem CID 23702152
SMILES [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O

Specifications

Concentration 0.2 M
pH 7.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water (350g/L) at 20°C,and methanol(very faint turbidity).
IUPAC Name sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
Chemical Name or Material TES buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only