missing translation for 'onlineSavingsMsg'
Learn More

tert-Pentylbenzene, 97%

Catalog No. AAL0373109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0373109 10 g
AAL0373118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0373109 Supplier Thermo Scientific Chemicals Supplier No. L0373109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2049-95-8 | C11H16 | 148.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2049-95-8
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00015187
InChI Key QHTJSSMHBLGUHV-UHFFFAOYSA-N
Synonym tert-amylbenzene, tert-pentylbenzene, 2-methyl-2-phenylbutane, 1,1-dimethylpropyl benzene, benzene, 1,1-dimethylpropyl, benzene, tert-pentyl, 2-phenyl-2-methylbutane, t-amylbenzene, 2-methylbutan-2-yl benzene, tert-amyl benzene
PubChem CID 16295
IUPAC Name 2-methylbutan-2-ylbenzene
SMILES CCC(C)(C)C1=CC=CC=C1

Specifications

Density 0.87
Boiling Point 188°C to 191°C
Flash Point 65°C (149°F)
Refractive Index 1.495
Quantity 10 g
Beilstein 2039053
Formula Weight 148.25
Percent Purity 97%
Chemical Name or Material tert-Pentylbenzene
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 218-076-0
RTECSNumber DA6720000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only