accessibility menu, dialog, popup

UserName

CAS RN 2049-95-8

IUPAC Name: (2-methylbutan-2-yl)benzene
Synonyms: benzene, (1,1-dimethylpropyl)-
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: QHTJSSMHBLGUHV-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)