missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylperoxy isopropyl carbonate, 75% solution in aromatic free mineral spirit

Catalog No. AC349842500
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC349842500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC349842500 Supplier Thermo Scientific Chemicals Supplier No. 349842500
Only null left

CAS: 2372-21-6 | C8H16O4 | 176.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2372-21-6
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.212
MDL Number MFCD00126878
InChI Key KDGNCLDCOVTOCS-UHFFFAOYSA-N
Synonym tert-butoxy isopropyl carbonate, carbonoperoxoic acid, oo-1,1-dimethylethyl o-1-methylethyl ester, o,o-tert-butyl isopropyl monoperoxycarbonate, peroxycarbonic acid, oo-tert-butyl isopropyl ester, 2-methyl-2-propan-2-yloxy carbonyl peroxy propane, carbonic acid tert-butoxy isopropyl ester, oo-tert-butyl=o-isopropyl=peroxycarbonate, oo-t-butyl o-isopropyl monoperoxycarbonate, tert-butylperoxy isopropyl carbonate solution, 2-isopropoxycarbonyl peroxy-2-methylpropane
PubChem CID 75406
IUPAC Name (2-methylpropan-2-yl)oxy propan-2-yl carbonate
SMILES CC(C)OC(=O)OOC(C)(C)C

Specifications

Linear Formula (CH3)2CHOCO3C(CH3)3
Formula Weight 176.21
Quantity 250 mL
Chemical Name or Material tert-Butylperoxy isopropyl carbonate, 75% solution in Aromatic Free Mineral Spirit

RUO – Research Use Only