accessibility menu, dialog, popup

UserName

H3CCH3OOOOCH3CH3H3C

CAS RN 2372-21-6

IUPAC Name: tert-butoxy propan-2-yl carbonate
Synonyms: tert-butoxy isopropyl carbonate
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: KDGNCLDCOVTOCS-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)OOC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results