missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydroquinone 98.0+%, TCI America™

Supplier:  TCI America B0833500G

 View more versions of this product

Catalog No. B0833500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

tert-Butylhydroquinone
127°C
C10H14O2
500 g
tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
CC(C)(C)C1=C(C=CC(=C1)O)O
166.22
CHEBI:78886
≥98.0% (GC)
1948-33-0
288°C
MFCD00002344
3077
BGNXCDMCOKJUMV-UHFFFAOYSA-N
2-tert-butylbenzene-1,4-diol
16043
166.22
Crystalline Powder

Chemical Identifiers

1948-33-0
166.22
BGNXCDMCOKJUMV-UHFFFAOYSA-N
16043
2-tert-butylbenzene-1,4-diol
C10H14O2
MFCD00002344
tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
CHEBI:78886
CC(C)(C)C1=C(C=CC(=C1)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0549

RTECSNumber : MX4375000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions