missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydroquinone, 97%

CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol

Supplier:  Thermo Scientific Chemicals A192060B

 View more versions of this product

Catalog No. AAA192060B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
TBHQ was used to study the inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli. Environment friendly electrode materials for supercapacitors were attained by decorating the surface of graphene nanosheets with TBHQ. TBHQ is frequently used as a food preservative. Inhibition of oxidative rancidity in frozen cooked fish flakes by tert-butylhydroquinone. Nuclear factor E2-related factor 2-dependent antioxidant response element activation is by tert-butylhydroquinoneoccurring preferentially in Astrocytes conditions.

Solubility
Slightly soluble in water(10g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

tert-Butylhydroquinone
126°C to 129°C
295°C
C10H14O2
1000 g
637923
Slightly soluble in water(10g/L).
CC(C)(C)C1=C(C=CC(=C1)O)O
166.22
CHEBI:78886
97%
1948-33-0
1.05
171°C (339°F)
MFCD00002344
UN3077
tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
BGNXCDMCOKJUMV-UHFFFAOYSA-N
2-tert-butylbenzene-1,4-diol
16043
166.22

Chemical Identifiers

1948-33-0
166.22
BGNXCDMCOKJUMV-UHFFFAOYSA-N
16043
2-tert-butylbenzene-1,4-diol
C10H14O2
MFCD00002344
tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
CHEBI:78886
CC(C)(C)C1=C(C=CC(=C1)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c

H302+H312-H315-H317-H319-H335-H341-H361

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 217-752-2

RTECSNumber : MX4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only