missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydroquinone, 97%

CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol

$70.03 - $651.17

Chemical Identifiers

CAS 1948-33-0
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002344
InChI Key BGNXCDMCOKJUMV-UHFFFAOYSA-N
Synonym tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
PubChem CID 16043
ChEBI CHEBI:78886
IUPAC Name 2-tert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=C(C=CC(=C1)O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC150820050
SDS View Documents
Thermo Scientific Chemicals
150820050
5 g Glass Bottle
Each for $70.03
Only null left
 
AC150822500
SDS View Documents
Thermo Scientific Chemicals
150822500
250 g Plastic bottle
Each for $116.55
Only null left
 
AC150825000
SDS View Documents
Thermo Scientific Chemicals
150825000
500 g Plastic bottle
Each for $190.97
Only null left
 
AC150820020
SDS View Documents
Thermo Scientific Chemicals
150820020
2 kg Plastic bottle
Each for $651.17
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1948-33-0
166.22
BGNXCDMCOKJUMV-UHFFFAOYSA-N
16043
2-tert-butylbenzene-1,4-diol
C10H14O2
MFCD00002344
tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
CHEBI:78886
CC(C)(C)C1=C(C=CC(=C1)O)O

Specifications

1948-33-0
Black-Tan to White
171°C
96% min. (GC)
C10H14O2
MFCD00002344
06, IV, 6013
Solubility in water: 1g/100mL (25 c). Other solubilities: soluble in acetone (190/100g at 25 c),,soluble in ethanol (120/100g at 25 c),,soluble in ethyl acetate (100g/100g at 25 c),,soluble in dmso,diethyl ether;,insoluble in benzene,toluene
CC(C)(C)C1=C(C=CC(=C1)O)O
166.22
CHEBI:78886
97%
tert-Butylhydroquinone, 97%
125.0°C to 130.0°C
295.0°C
Authentic
Glass Bottle
(CH3)3CC6H3(OH)2
5 g
tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
BGNXCDMCOKJUMV-UHFFFAOYSA-N
2-tert-butylbenzene-1,4-diol
16043
166.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-752-2

RTECSNumber : MX4375000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only