missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (Triphenylphosphoranylidene)carbamate 98.0+%, TCI America™

Catalog No. p-7124159
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56875G 5 g
B568725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56875G Supplier TCI America Supplier No. B56875G

Chemical Identifiers

CAS 68014-21-1
Molecular Formula C23H24NO2P
Molecular Weight (g/mol) 377.424
MDL Number MFCD00798174
InChI Key KNXPVXCUELYHDM-UHFFFAOYSA-N
Synonym (Triphenylphosphoranylidene)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)imino(triphenyl)phosphorane, N-Boc-imino(triphenyl)phosphorane
PubChem CID 2756802
IUPAC Name tert-butyl N-(triphenyl-$l^{5}-phosphanylidene)carbamate
SMILES CC(C)(C)OC(=O)N=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 149°C
Color White
Quantity 5 g
Formula Weight 377.42
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl (Triphenylphosphoranylidene)carbamate