accessibility menu, dialog, popup

UserName

H3CH3CCH3OONP

CAS RN 68014-21-1

IUPAC Name: tert-butyl N-(triphenyl-λ⁵-phosphanylidene)carbamate
Synonyms: tert-butyl N-(triphenyl-λ⁵-phosphanylidene)carbamate
Molecular Weight (g/mol): 377.42
Molecular Formula: C23H24NO2P
InChi Key: KNXPVXCUELYHDM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results