missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl bromoacetate, 98%

CAS: 5292-43-3 | C6H11BrO2 | 195.06 g/mol

Supplier:  Thermo Scientific Chemicals A1491718

 View more versions of this product

Catalog No. AAA1491718


Only null left
Explore available promotions

Description

Description

tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

tert-Butyl bromoacetate
1.33
49°C (120°F)
C6H11BrO2
BrCH2CO2C(CH3)3
50 g
1753010
Miscible with ethanol,chloroform and ethyl acetate. Immiscible with water.
CC(C)(C)OC(=O)CBr
195.06
195.06
5292-43-3
175°C
Pungent
1.445
MFCD00000188
UN2924
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat
BNWCETAHAJSBFG-UHFFFAOYSA-N
tert-butyl 2-bromoacetate
79177
98%

Chemical Identifiers

5292-43-3
195.06
BNWCETAHAJSBFG-UHFFFAOYSA-N
79177
CC(C)(C)OC(=O)CBr
C6H11BrO2
MFCD00000188
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat
tert-butyl 2-bromoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 226-133-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only