missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl bromoacetate, 98%

Catalog No. p-7022238
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1491709 10 g
AAA1491718 50 g
AAA1491730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1491709 Supplier Thermo Scientific Chemicals Supplier No. A1491709
Only null left

CAS: 5292-43-3 | C6H11BrO2 | 195.06 g/mol

tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 5292-43-3
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000188
InChI Key BNWCETAHAJSBFG-UHFFFAOYSA-N
Synonym tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat
PubChem CID 79177
IUPAC Name tert-butyl 2-bromoacetate
SMILES CC(C)(C)OC(=O)CBr

Specifications

Density 1.33
Boiling Point 175°C
Flash Point 49°C (120°F)
Odor Pungent
Refractive Index 1.445
Linear Formula BrCH2CO2C(CH3)3
Quantity 10 g
UN Number UN2924
Beilstein 1753010
Solubility Information Miscible with ethanol,chloroform and ethyl acetate. Immiscible with water.
Formula Weight 195.06
Percent Purity 98%
Chemical Name or Material tert-Butyl bromoacetate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 226-133-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only