missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate 95.0+%, TCI America™

Supplier:  TCI America B53851G

 View more versions of this product

Catalog No. B53851G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

tert-Butyl 6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Yellow
MFCD08691407
6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonyl)-3,4-epoxypyrrolidine, 1-Boc-3,4-epoxypyrrolidine
CC(C)(C)OC(=O)N1CC2C(C1)O2
185.223
185.22
Liquid
114214-49-2
C9H15NO3
1 g
NXZIGGBPLGAPTI-UHFFFAOYSA-N
tert-butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
10103880
≥95.0% (GC)

Chemical Identifiers

114214-49-2
185.223
NXZIGGBPLGAPTI-UHFFFAOYSA-N
10103880
CC(C)(C)OC(=O)N1CC2C(C1)O2
C9H15NO3
MFCD08691407
6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonyl)-3,4-epoxypyrrolidine, 1-Boc-3,4-epoxypyrrolidine
tert-butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions