accessibility menu, dialog, popup

UserName

CAS RN 114214-49-2

IUPAC Name: tert-butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Synonyms: tert-butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: NXZIGGBPLGAPTI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC2OC2C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results