missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate 95.0+%, TCI America™

Catalog No. p-7124063
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53851G 1 g
B53855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53851G Supplier TCI America Supplier No. B53851G

Chemical Identifiers

CAS 114214-49-2
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD08691407
InChI Key NXZIGGBPLGAPTI-UHFFFAOYSA-N
Synonym 6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonyl)-3,4-epoxypyrrolidine, 1-Boc-3,4-epoxypyrrolidine
PubChem CID 10103880
IUPAC Name tert-butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2C(C1)O2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 185.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate