Learn More
tert-Butyl 2-bromoisobutyrate, 97%
CAS: 23877-12-5 | C8H15BrO2 | 223.11 g/mol
Supplier: Thermo Scientific Chemicals L0821618
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| tert-Butyl 2-bromoisobutyrate | |
| 1.18 | |
| C8H15BrO2 | |
| MFCD00051908 | |
| 1905654 | |
| IGVNJALYNQVQIT-UHFFFAOYSA-N | |
| tert-butyl 2-bromo-2-methylpropanoate | |
| 90290 | |
| 97% |
| 23877-12-5 | |
| 35°C to 37°C (1 mmHg) | |
| 1.436 | |
| 50 g | |
| tert-butyl 2-bromoisobutyrate, t-butyl 2-bromo isobutyrate, 2-bromoisobutyric acid tert-butylester, tert-butyl 2-bromo-2-methylpropionate, tert-butyl alpha-bromoisobutyrate, t-butyl 2-bromoisobutylate, t-butyl 2-bromoisobutyrate, 2-bromoisobutyric acid tert-butyl ester, propanoic acid, 2-bromo-2-methyl-, 1,1-dimethylethyl ester, t-butyl2-bromoisobutyrate | |
| CC(C)(C)OC(=O)C(C)(C)Br | |
| 223.11 | |
| 223.12 |
Chemical Identifiers
| 23877-12-5 | |
| 223.11 | |
| IGVNJALYNQVQIT-UHFFFAOYSA-N | |
| tert-butyl 2-bromo-2-methylpropanoate |
| C8H15BrO2 | |
| MFCD00051908 | |
| 90290 | |
| CC(C)(C)OC(=O)C(C)(C)Br |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 245-924-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only